Anti-p-Hrs Rabbit Polyclonal Antibody [clone: Y216]
BWRLBS4824
- Antibody type:Primary
- Antigen symbol:p-Hrs
- Clonality:Polyclonal
- Clone:Y216
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic phosphopeptide derived from human Hrs around the phosphorylation site of Tyrosine 216.
p-Hrs (Y216) pAb detects endogenous levels of Hrs protein when phosphorylated at Tyr216
Type: Primary
Antigen: p-Hrs
Clonality: Polyclonal
Clone: Y216
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat