Anti-p-MEK1/2 Rabbit Polyclonal Antibody [clone: S218/222]
Catalog # BWRLBS4733
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-MEK1/2
- Clonality:Polyclonal
- Clone:S218/222
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human MEK1 around the phosphorylation site of Serine 218.
p-MEK1/2 (S218/222) pAb detects endogenous levels of MEK-1 protein only when phosphorylated at Ser218. This antibody also detects endogenous levels of MEK-2 protein only when phosphorylated at Thr222.
Type: Primary
Antigen: p-MEK1/2
Clonality: Polyclonal
Clone: S218/222
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat