Anti-ZNF682 Rabbit Polyclonal Antibody
Catalog # BWRLBS5981
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:ZNF682
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 61-106 of Human ZNF682.
ZNF682 pAb detects endogenous levels of ZNF682 protein.
Type: Primary
Antigen: ZNF682
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat