Anti-TOMM20 Rabbit Polyclonal Antibody
Catalog # BWRLBS5966
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:TOMM20
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 57-102 of Human TOMM20.
TOMM20 pAb detects endogenous levels of TOMM20 protein.
Type: Primary
Antigen: TOMM20
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat