Anti-p-MEK1 Rabbit Polyclonal Antibody [clone: T292]
Catalog # BWRLBS4709
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-MEK1
- Clonality:Polyclonal
- Clone:T292
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human MEK1 around the phosphorylation site of Threonine 292.
p-MEK1 (T292) pAb detects endogenous levels of MEK1protein only when phosphorylated at Thr292.
Type: Primary
Antigen: p-MEK1
Clonality: Polyclonal
Clone: T292
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat