Anti-SLC5A2 Rabbit Polyclonal Antibody
BWRLBS5929
- Antibody type:Primary
- Antigen symbol:Solute carrier family 5, member 2
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 151-196 of Human SLC5A2.
SLC5A2 pAb detects endogenous levels of SLC5A2 protein.
Type: Primary
Antigen: SLC5A2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat