Anti-PPP4C Rabbit Polyclonal Antibody
100 µg
Catalog # BWRLBS6703
Supplier: Bioworld Technology
Jump to:
Product Details & Documents
Recombinant full length Human PPP4C.
PPP4C pAb detects endogenous levels of PPP4C protein.
Type: Primary
Antigen: PPP4C
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Specifications
- Size:100 µg
- Western blot:Yes
- Cat. No.:BWRLBS6703
- Clonality:Polyclonal
- Reactivity:Human|Rat|Mouse
- Antibody type:Primary
- Host:Rabbit
- ImmunoChemistry:Yes
- Antigen symbol:PPP4C



