Order Entry
Export
ContactUsLinkComponent
Anti-CD95/FAS Rabbit Polyclonal Antibody
Anti-CD95/FAS Rabbit Polyclonal Antibody
  BWRLBS6430
Anti-CD95/FAS Rabbit Polyclonal Antibody
  BWRLBS6430

 

  • Antibody type:
    Primary
  • Antigen symbol:
    CD95/FAS
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Recombinant full length Human CD95/FAS.

Fas pAb detects endogenous levels of Fas protein.

Type: Primary
Antigen: CD95/FAS
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human