Order Entry
Export
ContactUsLinkComponent
Anti-Histone H1.0 Rabbit Polyclonal Antibody [clone: R94]
Anti-Histone H1.0 Rabbit Polyclonal Antibody [clone: R94]
Catalog # BWRLBS1916
Anti-Histone H1.0 Rabbit Polyclonal Antibody [clone: R94]
Catalog # BWRLBS1916

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Histone H1.0
  • Clonality:
    Polyclonal
  • Clone:
    R94
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to amino acids 61-110 of Human Histone H1.0.

Histone H1.0 (R94) pAb detects endogenous levels of Histone H1.0 protein.

Type: Primary
Antigen: Histone H1.0
Clonality: Polyclonal
Clone: R94
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human