Anti-TBX15/18 Rabbit Polyclonal Antibody [clone: D164]
Catalog # BWRLBS1907
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:TBX15/18
- Clonality:Polyclonal
- Clone:D164
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 131-180 of Human TBX15.
TBX15/18 (D164) pAb detects endogenous levels of TBX15 and TBX18 protein.
Type: Primary
Antigen: TBX15/18
Clonality: Polyclonal
Clone: D164
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse