Order Entry
Export
ContactUsLinkComponent
Anti-VEGI Rabbit Polyclonal Antibody [clone: F141]
Anti-VEGI Rabbit Polyclonal Antibody [clone: F141]
Catalog # BWRLBS1899
Anti-VEGI Rabbit Polyclonal Antibody [clone: F141]
Catalog # BWRLBS1899

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    VEGI
  • Clonality:
    Polyclonal
  • Clone:
    F141
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to amino acids 111-160 of Human VEGI.

VEGI (F141) pAb detects endogenous levels of VEGI protein.

Type: Primary
Antigen: VEGI
Clonality: Polyclonal
Clone: F141
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human