Anti-ALX3 Rabbit Polyclonal Antibody [clone: R203]
BWRLBS1898
- Antibody type:Primary
- Antigen symbol:ALX3
- Clonality:Polyclonal
- Clone:R203
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 171-220 of Human ALX3.
ALX3 (R203) pAb detects endogenous levels of ALX3 protein.
Type: Primary
Antigen: ALX3
Clonality: Polyclonal
Clone: R203
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat