Order Entry
Export
ContactUsLinkComponent
Anti-FIR Rabbit Polyclonal Antibody [clone: P364]
Anti-FIR Rabbit Polyclonal Antibody [clone: P364]
Catalog # BWRLBS1895
Anti-FIR Rabbit Polyclonal Antibody [clone: P364]
Catalog # BWRLBS1895

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    FIR
  • Clonality:
    Polyclonal
  • Clone:
    P364
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to amino acids 341-390 of Human FIR.

FIR (P364) pAb detects endogenous levels of FIR protein.

Type: Primary
Antigen: FIR
Clonality: Polyclonal
Clone: P364
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat