Anti-PRKY Rabbit Polyclonal Antibody [clone: H93]
BWRLBS3394
- Antibody type:Primary
- Antigen symbol:PRKY
- Clonality:Polyclonal
- Clone:H93
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 61-110 of Human PRKY.
PRKY (H93) pAb detects endogenous levels of PRKY protein.
Type: Primary
Antigen: PRKY
Clonality: Polyclonal
Clone: H93
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human