Anti-Vimentin Rabbit Polyclonal Antibody [clone: L79]
Catalog # BWRLBS1855
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Vimentin
- Clonality:Polyclonal
- Clone:L79
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 31-80 of Human Vimentin.
Vimentin (L79) pAb detects endogenous levels of Vimentin protein
Type: Primary
Antigen: Vimentin
Clonality: Polyclonal
Clone: L79
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat