Anti-GPR174 Rabbit Polyclonal Antibody [clone: K167]
Catalog # BWRLBS2779
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:GPR174
- Clonality:Polyclonal
- Clone:K167
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 132-178 of Human GPR174.
GPR174 (K167) pAb detects endogenous levels of GPR174 protein.
Type: Primary
Antigen: GPR174
Clonality: Polyclonal
Clone: K167
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat