Anti-Frizzled-7 Rabbit Polyclonal Antibody [clone: Y87]
BWRLBS2774
- Antibody type:Primary
- Antigen symbol:Frizzled-7
- Clonality:Polyclonal
- Clone:Y87
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 61-110 of Human Frizzled-7.
Frizzled-7 (Y87) pAb detects endogenous levels of Frizzled-7 protein.
Type: Primary
Antigen: Frizzled-7
Clonality: Polyclonal
Clone: Y87
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse