Anti-GLK Rabbit Polyclonal Antibody [clone: G4]
BWRLBS2755
- Antibody type:Primary
- Antigen symbol:GLK
- Clonality:Polyclonal
- Clone:G4
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to the N-terminal of Human GLK.
GLK (G4) pAb detects endogenous levels of GLK protein.
Type: Primary
Antigen: GLK
Clonality: Polyclonal
Clone: G4
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat