Anti-GPR153 Rabbit Polyclonal Antibody [clone: E234]
Catalog # BWRLBS2704
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:GPR153
- Clonality:Polyclonal
- Clone:E234
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 201-256 of Human GPR153.
GPR153 (E234) pAb detects endogenous levels of GPR153 protein.
Type: Primary
Antigen: GPR153
Clonality: Polyclonal
Clone: E234
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat