Anti-NDUFB10 Rabbit Polyclonal Antibody [clone: E95]
Catalog # BWRLBS3146
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:NDUFB10
- Clonality:Polyclonal
- Clone:E95
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 61-110 of Human NDUFB10.
NDUFB10 (E95) pAb detects endogenous levels of NDUFB10 protein.
Type: Primary
Antigen: NDUFB10
Clonality: Polyclonal
Clone: E95
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human