Anti-AURKB Rabbit Polyclonal Antibody [clone: Y8]
Catalog # BWRLBS1722
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:AURKB
- Clonality:Polyclonal
- Clone:Y8
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to the N-terminal of Human AURKB.
AURKB (Y8) pAb detects endogenous levels of AURKB protein
Type: Primary
Antigen: AURKB
Clonality: Polyclonal
Clone: Y8
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat