Anti-beta-1,4-Gal-T3 Rabbit Polyclonal Antibody [clone: F304]
Catalog # BWRLBS3090
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:beta-1,4-Gal-T3
- Clonality:Polyclonal
- Clone:F304
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 281-320 of Human beta-1,4-Gal-T3.
?-1,4-Gal-T3 (F304) pAb detects endogenous levels of?-1,4-Gal-T3 protein.
Type: Primary
Antigen: beta-1,4-Gal-T3
Clonality: Polyclonal
Clone: F304
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat