Anti-ApoL2 Rabbit Polyclonal Antibody [clone: R229]
BWRLBS3073
- Antibody type:Primary
- Antigen symbol:ApoL2
- Clonality:Polyclonal
- Clone:R229
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 196-240of Human ApoL2.
ApoL2 (R229) pAb detects endogenous levels of ApoL2 protein.
Type: Primary
Antigen: ApoL2
Clonality: Polyclonal
Clone: R229
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat