Order Entry
Export
ContactUsLinkComponent
Anti-LYPLA1 Rabbit Polyclonal Antibody [clone: K90]
Anti-LYPLA1 Rabbit Polyclonal Antibody [clone: K90]
Catalog # BWRLBS3063
Anti-LYPLA1 Rabbit Polyclonal Antibody [clone: K90]
Catalog # BWRLBS3063

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    LYPLA1
  • Clonality:
    Polyclonal
  • Clone:
    K90
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to amino acids 61-110 of Human LYPLA1.

LYPLA1 (K90) pAb detects endogenous levels of LYPLA1 protein.

Type: Primary
Antigen: LYPLA1
Clonality: Polyclonal
Clone: K90
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat