Anti-IFI-56K Rabbit Polyclonal Antibody [clone: H63]
Catalog # BWRLBS3033
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:IFI-56K
- Clonality:Polyclonal
- Clone:H63
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 31-80 of Human IFI-56K.
IFI-56K (H63) pAb detects endogenous levels of IFI-56K protein.
Type: Primary
Antigen: IFI-56K
Clonality: Polyclonal
Clone: H63
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human