Anti-GAL8 Rabbit Polyclonal Antibody [clone: N82]
BWRLBS2541
- Antibody type:Primary
- Antigen symbol:GAL8
- Clonality:Polyclonal
- Clone:N82
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 61-110 of Human Galectin-8.
Galectin-8 (N82) pAb detects endogenous levels of Galectin-8 protein.
Type: Primary
Antigen: GAL8
Clonality: Polyclonal
Clone: N82
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse