Anti-NIFK Rabbit Polyclonal Antibody [clone: D229]
BWRLBS1679
- Antibody type:Primary
- Antigen symbol:NIFK
- Clonality:Polyclonal
- Clone:D229
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 196-245 of Human NIFK.
NIFK (D229) pAb detects endogenous levels of NIFK protein.
Type: Primary
Antigen: NIFK
Clonality: Polyclonal
Clone: D229
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse