Anti-Ghrelin Rabbit Polyclonal Antibody [clone: K47]
100 µg
Catalog # BWRLBS2930
Supplier: Bioworld Technology
Anti-Ghrelin Rabbit Polyclonal Antibody [clone: K47]
Catalog # BWRLBS2930
Supplier: Bioworld Technology
Jump to:
Product Details & Documents
Synthetic peptide, corresponding to amino acids 16-64 of Human Ghrelin.
Ghrelin (K47) pAb detects endogenous levels of Ghrelin protein.
Type: Primary
Antigen: Ghrelin
Clonality: Polyclonal
Clone: K47
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Specifications
- Size:100 µg
- Western blot:Yes
- Cat. No.:BWRLBS2930
- Clonality:Polyclonal
- Reactivity:Human|Rat|Mouse
- Antibody type:Primary
- Host:Rabbit
- ImmunoChemistry:Yes
- Antigen symbol:Ghrelin
- Clone:K47



