Anti-PEA-15 Rabbit Polyclonal Antibody [clone: D110]
Catalog # BWRLBS1573
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:PEA-15
- Clonality:Polyclonal
- Clone:D110
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 78-132 of Human PEA-15.
PEA-15 (D110) pAb detects endogenous levels of PEA-15 protein.
Type: Primary
Antigen: PEA-15
Clonality: Polyclonal
Clone: D110
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat