Anti-CNOT4 Rabbit Polyclonal Antibody [clone: E95]
Catalog # BWRLBS3829
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:CNOT4
- Clonality:Polyclonal
- Clone:E95
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 61-110 of Human CNOT4.
CNOT4 (E95) pAb detects endogenous levels of CNOT4 protein.
Type: Primary
Antigen: CNOT4
Clonality: Polyclonal
Clone: E95
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat