Anti-Tsg 101 Rabbit Polyclonal Antibody [clone: E303]
Catalog # BWRLBS2347
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Tsg 101
- Clonality:Polyclonal
- Clone:E303
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 268-320 of Human Tsg 101.
Tsg 101 (E303) pAb detects endogenous levels of Tsg 101 protein.
Type: Primary
Antigen: Tsg 101
Clonality: Polyclonal
Clone: E303
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat