Anti-MEK1 Rabbit Polyclonal Antibody [clone: G281]
BWRLBS1549
- Antibody type:Primary
- Antigen symbol:MEK1
- Clonality:Polyclonal
- Clone:G281
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 260-310 of Human MEK1.
MEK1 (G281) pAb detects endogenous levels of MEK1 protein.
Type: Primary
Antigen: MEK1
Clonality: Polyclonal
Clone: G281
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat