Anti-Bcl-2 Rabbit Polyclonal Antibody [clone: P65]
Catalog # BWRLBS1511
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Bcl-2
- Clonality:Polyclonal
- Clone:P65
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 31-80 of Human Bcl-2.
Bcl-2 (P65) pAb detects endogenous levels of Bcl-2 protein.
Type: Primary
Antigen: Bcl-2
Clonality: Polyclonal
Clone: P65
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat