Anti-USF2 Rabbit Polyclonal Antibody [clone: D226]
Catalog # BWRLBS1489
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:USF2
- Clonality:Polyclonal
- Clone:D226
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 196-244 of Human USF2.
USF2 (D226) pAb detects endogenous levels of USF2 protein.
Type: Primary
Antigen: USF2
Clonality: Polyclonal
Clone: D226
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat