Anti-IL-16 Rabbit Polyclonal Antibody [clone: G97]
BWRLBS3690
- Antibody type:Primary
- Antigen symbol:IL-16
- Clonality:Polyclonal
- Clone:G97
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 66-122 of Human IL-16.
IL-16 (G97) pAb detects endogenous levels of IL-16 protein.
Type: Primary
Antigen: IL-16
Clonality: Polyclonal
Clone: G97
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human