Anti-Cardiotrophin-1 Rabbit Polyclonal Antibody [clone: H31]
BWRLBS3672
- Antibody type:Primary
- Antigen symbol:Cardiotrophin-1
- Clonality:Polyclonal
- Clone:H31
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 1-53 of Human CT-1.
Cardiotrophin-1 (H31) pAb detects endogenous levels of Cardiotrophin-1 protein.
Type: Primary
Antigen: Cardiotrophin-1
Clonality: Polyclonal
Clone: H31
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat