Anti-CPI-17 Rabbit Polyclonal Antibody [clone: K32]
Catalog # BWRLBS3643
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:CPI-17
- Clonality:Polyclonal
- Clone:K32
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 2-50 of Human CPI-17.
CPI-17 (K32) pAb pAb detects endogenous levels of CPI-17 pAb protein.
Type: Primary
Antigen: CPI-17
Clonality: Polyclonal
Clone: K32
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat