Anti-Emp Rabbit Polyclonal Antibody [clone: E207]
100 µg
Catalog # BWRLBS2092
Supplier: Bioworld Technology
Anti-Emp Rabbit Polyclonal Antibody [clone: E207]
Catalog # BWRLBS2092
Supplier: Bioworld Technology
Jump to:
Product Details & Documents
Synthetic peptide, corresponding to amino acids 178-228 of Human Emp.
Emp (E207) pAb detects endogenous levels of Emp protein.
Type: Primary
Antigen: Emp
Clonality: Polyclonal
Clone: E207
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Specifications
- Size:100 µg
- Western blot:Yes
- Cat. No.:BWRLBS2092
- Clonality:Polyclonal
- Reactivity:Human|Rat|Mouse
- Antibody type:Primary
- Host:Rabbit
- ImmunoChemistry:Yes
- Antigen symbol:Emp
- Clone:E207



