Order Entry
Export
ContactUsLinkComponent
Anti-p-ERalpha Rabbit Polyclonal Antibody [clone: S167]
Anti-p-ERalpha Rabbit Polyclonal Antibody [clone: S167]
Catalog # BWRLBS4072
Anti-p-ERalpha Rabbit Polyclonal Antibody [clone: S167]
Catalog # BWRLBS4072

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    p-ER alpha
  • Clonality:
    Polyclonal
  • Clone:
    S167
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic phosphopeptide derived from human ERalpha around the phosphorylation site of Serine 167.

p-ER? (S167) pAb detects endogenous levels of ER? protein only when phosphorylated at Ser167

Type: Primary
Antigen: p-ERalpha
Clonality: Polyclonal
Clone: S167
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat