Anti-p38 Rabbit Polyclonal Antibody [clone: H174]
Catalog # BWRLBS3566
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p38
- Clonality:Polyclonal
- Clone:H174
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 142-196 of Human p38.
p38 (H174) pAb detects endogenous levels of p38 protein.
Type: Primary
Antigen: p38
Clonality: Polyclonal
Clone: H174
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat