Anti-IL-11 Rabbit Polyclonal Antibody [clone: G27]
Catalog # BWRLBS3559
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:IL-11
- Clonality:Polyclonal
- Clone:G27
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to the N-terminus of Human IL-11.
IL-11 (G27) pAb detects endogenous levels of IL-11 protein.
Type: Primary
Antigen: IL-11
Clonality: Polyclonal
Clone: G27
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat