Anti-GCKR Rabbit Polyclonal Antibody [clone: R227]
BWRLBS2059
- Antibody type:Primary
- Antigen symbol:GCKR
- Clonality:Polyclonal
- Clone:R227
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 196-245 of Human GCKR.
GCKR (R227) pAb detects endogenous levels of GCKR protein.
Type: Primary
Antigen: GCKR
Clonality: Polyclonal
Clone: R227
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat