Anti-NPY Rabbit Polyclonal Antibody [clone: P94]
BWRLBS3551
- Antibody type:Primary
- Antigen symbol:NPY
- Clonality:Polyclonal
- Clone:P94
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 61-110 of Human NPY.
NPY (P94) pAb detects endogenous levels of NPY protein.
Type: Primary
Antigen: NPY
Clonality: Polyclonal
Clone: P94
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat