Order Entry
Export
Orders LinkContactUsLinkComponent
 

Anti-EphB4 Rabbit Polyclonal Antibody [clone: E601]

100 µg
  BWRLBS2018
Anti-EphB4 Rabbit Polyclonal Antibody [clone: E601]
  BWRLBS2018
Anti-EphB4 Rabbit Polyclonal Antibody [clone: E601]

100 µg

 
250,00 €
   
1 * 100 µG
 

Synthetic peptide, corresponding to amino acids 580-630 of Human EphB4.

EphB4 (E601) pAb detects endogenous levels of EphB4 protein.

Type: Primary
Antigen: EphB4
Clonality: Polyclonal
Clone: E601
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat

 
  • Size:
    100 µg
  • Western blot:
    Yes
  • Cat. No.:
    BWRLBS2018
  • Clonality:
    Polyclonal
  • Reactivity:
    Human|Rat|Mouse
  • Antibody type:
    Primary
  • ImmunoFluorescence:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Antigen symbol:
    EphB4
  • Clone:
    E601