Anti-GDF-15 Rabbit Polyclonal Antibody [clone: L64]
Catalog # BWRLBS3872
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:GDF-15
- Clonality:Polyclonal
- Clone:L64
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 32-80 of Human GDF-15.
GDF-15 (L64) pAb detects endogenous levels of GDF-15 protein.
Type: Primary
Antigen: GDF-15
Clonality: Polyclonal
Clone: L64
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat