Anti-IFN-gamma Rabbit Polyclonal Antibody [clone: Q69]
BWRLBS3486
- Antibody type:Primary
- Antigen symbol:IFN- gamma
- Clonality:Polyclonal
- Clone:Q69
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 31-80 of Human IFN-gamma.
IFN-? (Q69) pAb detects endogenous levels of IFN-? protein.
Type: Primary
Antigen: IFN-gamma
Clonality: Polyclonal
Clone: Q69
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human