Order Entry
Export
ContactUsLinkComponent
Anti-CD3-epsilon Rabbit Polyclonal Antibody [clone: D71]
Anti-CD3-epsilon Rabbit Polyclonal Antibody [clone: D71]
Catalog # BWRLBS3476
Anti-CD3-epsilon Rabbit Polyclonal Antibody [clone: D71]
Catalog # BWRLBS3476

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    CD3- epsilon
  • Clonality:
    Polyclonal
  • Clone:
    D71
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to amino acids 31-80 of Human CD3-epsilon.

CD3-? (D71) pAb detects endogenous levels of CD3-? protein.

Type: Primary
Antigen: CD3-epsilon
Clonality: Polyclonal
Clone: D71
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat