Anti-IL-5 Rabbit Polyclonal Antibody [clone: S92]
Catalog # BWRLBS3471
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:IL-5
- Clonality:Polyclonal
- Clone:S92
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 61-110 of Human IL-5.
IL-5 (S92) pAb detects endogenous levels of IL-5 protein.
Type: Primary
Antigen: IL-5
Clonality: Polyclonal
Clone: S92
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat