Anti-CNPase Rabbit Polyclonal Antibody [clone: A108]
Catalog # BWRLBS3461
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:CNPase
- Clonality:Polyclonal
- Clone:A108
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 61-110 of Human CNPase.
CNPase (A108) pAb detects endogenous levels of CNPase protein.
Type: Primary
Antigen: CNPase
Clonality: Polyclonal
Clone: A108
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat