Anti-CD44 Rabbit Polyclonal Antibody [clone: E702]
100 µg
Catalog # BWRLBS3431
Supplier: Bioworld Technology
Anti-CD44 Rabbit Polyclonal Antibody [clone: E702]
Catalog # BWRLBS3431
Supplier: Bioworld Technology
Jump to:
Product Details & Documents
Synthetic peptide, corresponding to amino acids 670-720 of Human CD44.
CD44 (E702) pAb detects endogenous levels of CD44 protein.
Type: Primary
Antigen: CD44
Clonality: Polyclonal
Clone: E702
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Specifications
- Size:100 µg
- Western blot:Yes
- Cat. No.:BWRLBS3431
- Clonality:Polyclonal
- Reactivity:Human|Rat|Mouse
- Antibody type:Primary
- Host:Rabbit
- Antigen symbol:CD44
- Clone:E702



